Learn More
Cayman Chemical Lauric Acid-d23, Purity: ≥99% deuterated forms (d1-d23), CAS: 59154-43-7, 10 mg
SDP
Catalog No. 502937376
Encompass_Preferred
An internal standard for the quantification of lauric acid by GC- or LC-MS
Catalog No. 50-293-7376 Supplier Cayman Chemical Supplier No. 2808010 MG
An internal standard for the quantification of lauric acid by GC- or LC-MS