Learn More
Cayman Chemical Lauric Acid-d23, Purity: ≥99% deuterated forms (d1-d23), CAS: 59154-43-7, 50 mg
SDP
Catalog No. 502939960
Encompass_Preferred
An internal standard for the quantification of lauric acid by GC- or LC-MS
Catalog No. 50-293-9960 Supplier Cayman Chemical Supplier No. 2808050 MG
An internal standard for the quantification of lauric acid by GC- or LC-MS