Learn More
Medchemexpress LLC Lauric acid-d23 | 59154-43-7 | MFCD00144567 | 98.4% | 223.46 g·mol⁻¹ | C12HD23O2 | 25mg
SDP
Supplier:  Medchemexpress LLC HYY0366S125MG
Encompass_Preferred
Lauric acid-d233 is the deuterium labeled Lauric acid Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively
Catalog No. 50-002-68277