Learn More
Cayman Chemical Lauric Acid ethyl ester, Purity: ≥98%, CAS: 106-33-2, 100 mg
SDP
Catalog No. 502888682
Encompass_Preferred
Lauric acid ethyl ester is a more lipophilic and less toxic form of the free acid. It is one of the medium-chain fatty acid ethyl esters that is released during the anaerobic fermentation of Saccharomyces cerevisiae along with the free acid.
Catalog No. 50-288-8682 Supplier Cayman Chemical Supplier No. 10008203100 MG
Lauric acid ethyl ester is a more lipophilic and less toxic form of the free acid. It is one of the medium-chain fatty acid ethyl esters that is released during the anaerobic fermentation of Saccharomyces cerevisiae along with the free acid.