Learn More
Cayman Chemical Lauric Acid, Purity: ≥98%, CAS: 143-07-7, 100 g
SDP
Catalog No. 502888137
Encompass_Preferred
A medium-chain fatty acid; has been found at high levels in coconut oil; induces the activation of NF-κB and the expression of COX-2, iNOS, and IL-1α in RAW 264.7 cells at 25 µM
Catalog No. 50-288-8137 Supplier Cayman Chemical Supplier No. 10006626100 G
A medium-chain fatty acid; has been found at high levels in coconut oil; induces the activation of NF-κB and the expression of COX-2, iNOS, and IL-1α in RAW 264.7 cells at 25 µM