Learn More
Cayman Chemical Lauric Acid Leelamide, Purity: ≥98%, CAS: 2095306-47-9, 5 mg
SDP
Catalog No. 502891435
Encompass_Preferred
Lauric acid leelamide is the lauric (C-12) amide analog of leelamine. Leelamine has weak affinity for the human CB1 and CB2 receptors, exhibiting 20% displacement of [3H]-CP55940 at a concentration of 10 µM.{14127} Leelamine inhibits PDHK with an IC50 value of 9.5 µM.{14105} Derivatives of leelamine exhibit anti-inflammatory activity and show moderate inhibition of PLA2 activity from a variety of sources.{14107} There are no published studies of the pharmacological properties of lauric acid leelamide.
Catalog No. 50-289-1435 Supplier Cayman Chemical Supplier No. 100086185 MG
Lauric acid leelamide is the lauric (C-12) amide analog of leelamine. Leelamine has weak affinity for the human CB1 and CB2 receptors, exhibiting 20% displacement of [3H]-CP55940 at a concentration of 10 µM.{14127} Leelamine inhibits PDHK with an IC50 value of 9.5 µM.{14105} Derivatives of leelamine exhibit anti-inflammatory activity and show moderate inhibition of PLA2 activity from a variety of sources.{14107} There are no published studies of the pharmacological properties of lauric acid leelamide.