Learn More
Cayman Chemical Lauric Acid methyl ester, Purity: ≥98%, CAS: 111-82-0, 500 mg
SDP
Catalog No. 502984845
Encompass_Preferred
An esterified version of lauric acid; a major component in a biodiesel preparation made from crude fat extracted from black soldier flies; used as an internal standard for GC- and LC-MS
Catalog No. 50-298-4845 Supplier Cayman Chemical Supplier No. 20608500 MG
An esterified version of lauric acid; a major component in a biodiesel preparation made from crude fat extracted from black soldier flies; used as an internal standard for GC- and LC-MS