Learn More
Medchemexpress LLC Lauric anhydride 10g
SDP
Catalog No. 5000206886
Encompass_Preferred
Lauric anhydride is a white waxy solid used as an intermediate in the production of various chemicals It is primarily used in the synthesis of surfactants emulsifiers and lubricants and as a raw material for other important industrial chemicals such as plasticizers and flavoring agents It can be obtained from natural sources such as palm kernel oil and coconut oil and its unique chemical structure allows it to be used to create compounds with specific surface and interfacial properties
Catalog No. 50-002-06886 Supplier Medchemexpress LLC Supplier No. HYW09960010G
Lauric anhydride is a white waxy solid used as an intermediate in the production of various chemicals It is primarily used in the synthesis of surfactants emulsifiers and lubricants and as a raw material for other important industrial chemicals such as plasticizers and flavoring agents It can be obtained from natural sources such as palm kernel oil and coconut oil and its unique chemical structure allows it to be used to create compounds with specific surface and interfacial properties