Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lauric Anhydride, MP Biomedicals

Catalog No. ICN215501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN215501 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN215501 Supplier MP Biomedicals, Inc Supplier No. 0521550105
Add to Cart
Add to Cart

Chemical Identifiers

CAS 645-66-9
Molecular Formula C24H46O3
Molecular Weight (g/mol) 382.629
InChI Key NWADXBLMWHFGGU-UHFFFAOYSA-N
Synonym lauric anhydride, dodecanoic anhydride, lauroyl anhydride, dodecanoic acid, anhydride, n-dodecanoic anhydride, lauricanhydride, dodecanoyl anhydride, lauric acid anhydride, dodecanoic anhydride #, dodecanoicacid,anhydride
PubChem CID 69527
IUPAC Name dodecanoyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC

Specifications

Quantity 10 g
Chemical Name or Material Lauric Anhydride
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.