Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lauric Anhydride 98.0+%, TCI America™
SDP

Catalog No. L0017100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0017100G 100 g
L001725G 25 g
L0017500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L0017100G Supplier TCI America Supplier No. L0017100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 645-66-9
Molecular Formula C24H46O3
Molecular Weight (g/mol) 382.629
MDL Number MFCD00008965
InChI Key NWADXBLMWHFGGU-UHFFFAOYSA-N
Synonym lauric anhydride, dodecanoic anhydride, lauroyl anhydride, dodecanoic acid, anhydride, n-dodecanoic anhydride, lauricanhydride, dodecanoyl anhydride, lauric acid anhydride, dodecanoic anhydride #, dodecanoicacid,anhydride
PubChem CID 69527
IUPAC Name dodecanoyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC

Specifications

Melting Point 42°C
Color White
Quantity 100 g
Formula Weight 382.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lauric Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.