Learn More
Medchemexpress LLC Laurocapram | 59227-89-3 | 281.48 | 1 ML
SDP
Catalog No. 5000366851
Encompass_Preferred
Laurocapram is a chemical penetration enhancer that increases the permeability of compounds through the skin and mucous membranes, improving their local or systemic efficacy. It enhances the skin permeability of various hydrophilic and lipophilic compounds. This product is provided as a 10 mM solution in 1 mL, suitable for research applications.
  • Increases permeability of compounds through skin and mucous membranes.
  • Improves local or systemic efficacy of compounds.
  • Enhances skin permeability for both hydrophilic and lipophilic compounds.
  • Synergizes with β-lactam antibiotics to improve antibacterial activity against S. aureus.
  • Exhibits anti-infectious activity in MRSA-induced mouse pneumonia models.
  • Ameliorates infection wounds in MRSA-administrated mouse skin infection models.
Catalog No. 50-003-66851 Supplier Medchemexpress LLC Supplier No. HYW00932610MM/1ML
Laurocapram is a chemical penetration enhancer that increases the permeability of compounds through the skin and mucous membranes, improving their local or systemic efficacy. It enhances the skin permeability of various hydrophilic and lipophilic compounds. This product is provided as a 10 mM solution in 1 mL, suitable for research applications.
  • Increases permeability of compounds through skin and mucous membranes.
  • Improves local or systemic efficacy of compounds.
  • Enhances skin permeability for both hydrophilic and lipophilic compounds.
  • Synergizes with β-lactam antibiotics to improve antibacterial activity against S. aureus.
  • Exhibits anti-infectious activity in MRSA-induced mouse pneumonia models.
  • Ameliorates infection wounds in MRSA-administrated mouse skin infection models.