Learn More
Apexbio Technology LLC Laurocapram 59227-89-3 500mg
SDP
Catalog No. 5000089705
Encompass_Preferred
Laurocapram is an amphiphilic compound primarily applied as a percutaneous absorption enhancer Laurocapram exerts its biological activity by temporarily altering the lipid architecture within the stratum corneum thereby lowering barrier resistance and increasing skin and mucosal permeability to both hydrophilic and lipophilic substances Based on these pharmacological properties Laurocapram holds research potential in the investigation of transdermal drug delivery mechanisms evaluation of pharmacokinetic profiles and the development of topical therapeutic agents and delivery formulations
Catalog No. 50-000-89705 Supplier Apexbio Technology LLC Supplier No. C6524500
Laurocapram is an amphiphilic compound primarily applied as a percutaneous absorption enhancer Laurocapram exerts its biological activity by temporarily altering the lipid architecture within the stratum corneum thereby lowering barrier resistance and increasing skin and mucosal permeability to both hydrophilic and lipophilic substances Based on these pharmacological properties Laurocapram holds research potential in the investigation of transdermal drug delivery mechanisms evaluation of pharmacokinetic profiles and the development of topical therapeutic agents and delivery formulations