Learn More
Medchemexpress LLC Laurocapram | 59227-89-3 | 99.4% | 281.48 | 5 G
SDP
Catalog No. 5000451671
Encompass_Preferred
Laurocapram is a chemical penetration enhancer that increases the permeability of compounds through the skin and mucous membranes, improving their local or systemic efficacy. It enhances the skin permeability of various hydrophilic and lipophilic compounds.
  • Synergizes with β-lactam antibiotics like Cefixime, Penicillin G, and Polymyxin E to improve antibacterial activity against S. aureus, especially MRSA strains.
  • Exhibits anti-infectious activity in MRSA-induced mouse pneumonia models.
  • Ameliorates infection wounds in MRSA-administered mouse skin infection models.
  • Alleviates pathological changes of lung tissue.
  • Reduces pulmonary edema and bacterial load.
  • Reduces skin infection wound area.
  • Decreases levels of FN-γ, IL-6, and TNF-α.
Catalog No. 50-004-51671 Supplier Medchemexpress LLC Supplier No. HYW0093265G
Laurocapram is a chemical penetration enhancer that increases the permeability of compounds through the skin and mucous membranes, improving their local or systemic efficacy. It enhances the skin permeability of various hydrophilic and lipophilic compounds.
  • Synergizes with β-lactam antibiotics like Cefixime, Penicillin G, and Polymyxin E to improve antibacterial activity against S. aureus, especially MRSA strains.
  • Exhibits anti-infectious activity in MRSA-induced mouse pneumonia models.
  • Ameliorates infection wounds in MRSA-administered mouse skin infection models.
  • Alleviates pathological changes of lung tissue.
  • Reduces pulmonary edema and bacterial load.
  • Reduces skin infection wound area.
  • Decreases levels of FN-γ, IL-6, and TNF-α.