Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Laurolitsine (hydrochloride) | 25 MG
SDP

Catalog No. 5000421908
Encompass_Preferred

Laurolitsine hydrochloride is an alkaloid isolated from the leaves of Peumus boldus Molina. It demonstrates weak anti-inflammatory activity and inhibits aggregation of rabbit platelets.

  • Purity of 99.97%
  • Appears as a solid, white to pink
  • Recommended storage at 4°C in sealed conditions, away from moisture
  • For storage in solvent, -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
  • Solubility of 100 mg/mL in DMSO

Catalog No. 50-004-21908 Supplier Medchemexpress LLC Supplier No. 25MGHYN2352A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laurolitsine hydrochloride is an alkaloid isolated from the leaves of Peumus boldus Molina. It demonstrates weak anti-inflammatory activity and inhibits aggregation of rabbit platelets.

  • Purity of 99.97%
  • Appears as a solid, white to pink
  • Recommended storage at 4°C in sealed conditions, away from moisture
  • For storage in solvent, -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
  • Solubility of 100 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.