Learn More
Medchemexpress LLC Laurolitsine (hydrochloride) | 50 MG
SDP
Catalog No. 5000421063
Encompass_Preferred
Laurolitsine hydrochloride, also known as (+)-Norboldine hydrochloride, is an alkaloid derived from the leaves of *Peumus boldus* Molina. This compound exhibits weak anti-inflammatory activity and is intended for research use only. It has been shown to inhibit NO production in RAW 267.4 and BV-2 cells, and also remarkably inhibits the aggregation of rabbit platelets.
  • Alkaloid isolated from *Peumus boldus* Molina leaves
  • Exhibits weak anti-inflammatory activity
  • Inhibits NO production in RAW 267.4 and BV-2 cells
  • Inhibits aggregation of rabbit platelets
Catalog No. 50-004-21063 Supplier Medchemexpress LLC Supplier No. HYN2352A50MG
Laurolitsine hydrochloride, also known as (+)-Norboldine hydrochloride, is an alkaloid derived from the leaves of *Peumus boldus* Molina. This compound exhibits weak anti-inflammatory activity and is intended for research use only. It has been shown to inhibit NO production in RAW 267.4 and BV-2 cells, and also remarkably inhibits the aggregation of rabbit platelets.
  • Alkaloid isolated from *Peumus boldus* Molina leaves
  • Exhibits weak anti-inflammatory activity
  • Inhibits NO production in RAW 267.4 and BV-2 cells
  • Inhibits aggregation of rabbit platelets