Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Laurolitsine hydrochloride | 5890-18-6 | MFCD06809897 | 100.0% | 349.81 | C18H20ClNO4 | 10 MG
SDP

Catalog No. 5000289614
Encompass_Preferred

Laurolitsine hydrochloride is a naturally occurring aporphine alkaloid isolated from Phoebe formosana and supplied for research use. It has been reported to exhibit weak anti-inflammatory activity in cell-based assays and shows activity in platelet aggregation studies.

  • Natural aporphine alkaloid isolated from Phoebe formosana.
  • Shows weak anti-inflammatory activity in cell assays.
  • Chemical formula C18H20ClNO4; molecular weight 349.81.
  • Purity 99.97% as supplied.
  • Solid, off-white to pink appearance.
  • Storage: 4 °C sealed, away from moisture; in solvent -80 °C (6 months), -20 °C (1 month).

Catalog No. 50-002-89614 Supplier Medchemexpress LLC Supplier No. HYN2352A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laurolitsine hydrochloride is a naturally occurring aporphine alkaloid isolated from Phoebe formosana and supplied for research use. It has been reported to exhibit weak anti-inflammatory activity in cell-based assays and shows activity in platelet aggregation studies.

  • Natural aporphine alkaloid isolated from Phoebe formosana.
  • Shows weak anti-inflammatory activity in cell assays.
  • Chemical formula C18H20ClNO4; molecular weight 349.81.
  • Purity 99.97% as supplied.
  • Solid, off-white to pink appearance.
  • Storage: 4 °C sealed, away from moisture; in solvent -80 °C (6 months), -20 °C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.