Learn More
American Radiolabeled Chemicals Inc LAUROYL-1-14C COA 50UC
SDP
Catalog No. 5090103713
Encompass_Preferred
Lauroyl-1-14C Coenzyme A 50uCi in 0.1M sodium acetate buffer (pH 5.2), 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 949.8, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-03713 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 083250UC
Add to Cart
Add to Cart
Request a Quote
Lauroyl-1-14C Coenzyme A 50uCi in 0.1M sodium acetate buffer (pH 5.2), 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 949.8, Shipped in Dry Ice, Exclusive