Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lauroyl chloride

Catalog No. 1569301L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-693-01L 1 L
15-693-0250ML 250 mL
15-693-050ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-693-01L Supplier Sigma Aldrich Supplier No. 1569301L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Lauroyl chloride was used:
  • As tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals
  • In the preparation of acylated collagen with water solubility and better surface activity
  • As organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel
  • In the synthesis of hemicellulose-based hydrophobic biomaterials

Specifications

CAS 112-16-3
Density 0.946 g/mL (at 25°C (literature))
Boiling Point 134°C to 137°C (11 mmHg)
Molecular Formula C12H23ClO
Refractive Index n20/D 1.445 (literature)
Linear Formula CH3(CH2)10 COCl
MDL Number MFCD00000740
Quantity 1 L
Synonym Dodecanoyl chloride
Molecular Weight (g/mol) 218.76
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.