Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lauroyl coenzyme A (lithium) | 190063-12-8 | 99% | 1 MG
SDP

Catalog No. 5000343012
Encompass_Preferred

Lauroyl coenzyme A lithium salt is an intermediary in fatty acid synthesis or metabolism. It is formed by combining long-chain fatty acids (specifically lauric acid) with coenzyme A. This compound plays a role in lipid biosynthesis and fatty acid transport, where coenzyme A acts as a transport molecule to facilitate the movement and targeting of specific compounds.

  • Intermediary in fatty acid synthesis or metabolism.
  • Formed by combining long-chain fatty acids (lauric acid) with coenzyme A.
  • Involved in lipid biosynthesis and fatty acid transport.
  • Coenzyme A acts as a transport molecule to help move and target specific compounds.
  • Purity: 99%.
  • Appearance: White to off-white solid.

Catalog No. 50-003-43012 Supplier Medchemexpress LLC Supplier No. HY126912A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lauroyl coenzyme A lithium salt is an intermediary in fatty acid synthesis or metabolism. It is formed by combining long-chain fatty acids (specifically lauric acid) with coenzyme A. This compound plays a role in lipid biosynthesis and fatty acid transport, where coenzyme A acts as a transport molecule to facilitate the movement and targeting of specific compounds.

  • Intermediary in fatty acid synthesis or metabolism.
  • Formed by combining long-chain fatty acids (lauric acid) with coenzyme A.
  • Involved in lipid biosynthesis and fatty acid transport.
  • Coenzyme A acts as a transport molecule to help move and target specific compounds.
  • Purity: 99%.
  • Appearance: White to off-white solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.