Learn More
Medchemexpress LLC Lauroyl-L-carnitine (chloride) | 6919-91-1 | MFCD00274131 | 98.1% | 379.96 g·mol⁻¹ | C19H38ClNO4 | 5 MG
SDP
Catalog No. 5000263166
Encompass_Preferred
Lauroyl-L-carnitine chloride is an acylcarnitine-derived absorption enhancer used in research to increase the oral bioavailability of peptide compounds. It is provided as a high-purity solid for laboratory use and can be prepared as solutions for experimental assays following recommended storage conditions.
  • Orally active absorption enhancer for peptide compounds.
  • Improves oral bioavailability of peptide therapeutics in preclinical studies.
  • Supplied as a high-purity solid suitable for solution preparation.
  • Available in small research pack sizes and larger quantities on request.
  • Stable when stored sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • Molecular formula C19H38ClNO4; molecular weight 379.96 g·mol⁻¹.
Catalog No. 50-002-63166 Supplier Medchemexpress LLC Supplier No. HY1303215MG
Lauroyl-L-carnitine chloride is an acylcarnitine-derived absorption enhancer used in research to increase the oral bioavailability of peptide compounds. It is provided as a high-purity solid for laboratory use and can be prepared as solutions for experimental assays following recommended storage conditions.
  • Orally active absorption enhancer for peptide compounds.
  • Improves oral bioavailability of peptide therapeutics in preclinical studies.
  • Supplied as a high-purity solid suitable for solution preparation.
  • Available in small research pack sizes and larger quantities on request.
  • Stable when stored sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • Molecular formula C19H38ClNO4; molecular weight 379.96 g·mol⁻¹.