Learn More
Medchemexpress LLC Lauroyl-L-carnitine chloride | 6919-91-1 | MFCD00274131 | 98.1% | C19H38ClNO4 | 10MG
SDP
Catalog No. 5000260930
Encompass_Preferred
Lauroyl-L-carnitine chloride is an orally active absorption enhancer used in research to improve the oral bioavailability of peptide compounds. Supplied as a white to off-white solid with high purity, it is applied in formulation and permeability studies to assess and enhance intestinal uptake of peptide drugs.
  • Orally active absorption enhancer for peptide compounds.
  • Reported to improve oral bioavailability of peptides.
  • White to off-white solid suitable for formulation research.
  • High purity (~98.1%) as supplied.
  • Supports studies of intestinal permeability and delivery.
  • Store sealed at 4°C; in solvent, -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-60930 Supplier Medchemexpress LLC Supplier No. HY13032110MG
Lauroyl-L-carnitine chloride is an orally active absorption enhancer used in research to improve the oral bioavailability of peptide compounds. Supplied as a white to off-white solid with high purity, it is applied in formulation and permeability studies to assess and enhance intestinal uptake of peptide drugs.
  • Orally active absorption enhancer for peptide compounds.
  • Reported to improve oral bioavailability of peptides.
  • White to off-white solid suitable for formulation research.
  • High purity (~98.1%) as supplied.
  • Supports studies of intestinal permeability and delivery.
  • Store sealed at 4°C; in solvent, -80°C (6 months) or -20°C (1 month).