Learn More
Medchemexpress LLC Lauroyl-L-carnitine-d9 chloride | 2245711-25-3 | MFCD34676545 | 98.0% | 389.02 g/mol | C19H29D9ClNO4 | 5 MG
SDP
Catalog No. 5000263167
Encompass_Preferred
Lauroyl-L-carnitine-d9 chloride is the deuterium-labeled chloride salt of lauroyl-L-carnitine supplied for analytical and tracer applications. It is intended for use as a stable-isotope internal standard in bioanalytical, pharmacokinetic, and tracer studies where mass spectrometry differentiation is required.
  • Stable isotope labeled internal standard for mass spectrometry
  • Deuterated (d9) for clear mass separation from native compound
  • High purity (98.0%) suitable for analytical workflows
  • Chloride salt form for compatibility with common assays
  • Supplied in a 5 mg quantity for small-scale analyses
Catalog No. 50-002-63167 Supplier Medchemexpress LLC Supplier No. HY130321S15MG
Lauroyl-L-carnitine-d9 chloride is the deuterium-labeled chloride salt of lauroyl-L-carnitine supplied for analytical and tracer applications. It is intended for use as a stable-isotope internal standard in bioanalytical, pharmacokinetic, and tracer studies where mass spectrometry differentiation is required.
  • Stable isotope labeled internal standard for mass spectrometry
  • Deuterated (d9) for clear mass separation from native compound
  • High purity (98.0%) suitable for analytical workflows
  • Chloride salt form for compatibility with common assays
  • Supplied in a 5 mg quantity for small-scale analyses