Learn More
Medchemexpress LLC Lauroyl lysine 10g | 52315-75-0 | 10 G

Supplier: Medchemexpress LLC HY11719510G
Lauroyl lysine (L-lysine, N6-(1-oxododecyl)-; CAS 52315-75-0) is an amino acid derivative supplied as a white to off-white powder for research and formulation use. It has a molecular weight of 328.49 g/mol and is commonly used in cosmetic formulations to improve skin feel and as a biochemical reagent in laboratory studies.
- Purity ≥98.0%.
- Molecular formula C18H36N2O3.
- Available as a 10 G powder pack for laboratory use.
- White to off-white powder physical form.
- Suitable for formulation and analytical research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.