Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lauroylcholine Chloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. D09731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09731G 1 g
D09735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D09731G Supplier TCI America Supplier No. D09731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25234-60-0
Molecular Formula C17H36ClNO2
Molecular Weight (g/mol) 321.93
MDL Number MFCD00050185
InChI Key BBJUKVPDIPYNBS-UHFFFAOYSA-M
Synonym Dodecanoylcholine Chloride
PubChem CID 91343
IUPAC Name 2-dodecanoyloxyethyl(trimethyl)azanium;chloride
SMILES CCCCCCCCCCCC(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

Color White
Quantity 1 g
Formula Weight 321.93
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lauroylcholine Chloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.