Learn More
Cayman Chemical Lauroyl-DL-Carnitine (chloride), Purity: ≥95%, CAS: 14919-37-0, 50 mg
SDP
Catalog No. 502968986
Encompass_Preferred
A zwitterionic, long-chain acylcarnitine used to improve in vivo absorption of certain hydrophilic compounds, especially through mucosal membranes
Catalog No. 50-296-8986 Supplier Cayman Chemical Supplier No. 1600650 MG
A zwitterionic, long-chain acylcarnitine used to improve in vivo absorption of certain hydrophilic compounds, especially through mucosal membranes