Learn More
Cayman Chemical Lauroyl-L-carnitine (chloride), Purity: ≥98%, CAS: 6919-91-1, 50 mg
SDP
Catalog No. 502933638
Encompass_Preferred
An acylcarnitine and a quaternary ammonium-containing cationic surfactant; has been used to permeabilize porcine enterocytes for delivery of the polar fluorescent probe lucifer yellow
Catalog No. 50-293-3638 Supplier Cayman Chemical Supplier No. 2655050 MG
An acylcarnitine and a quaternary ammonium-containing cationic surfactant; has been used to permeabilize porcine enterocytes for delivery of the polar fluorescent probe lucifer yellow