Learn More
Crescent Chemical Co Inc LAUROYLSARCOSINE-NA-SALT 30%
SDP
Supplier:  Crescent Chemical Co Inc SE27570.02
Encompass_Preferred
Lauroylsarcosine-Na-salt 30 % solution Disinfectant. Active substance 28.5 - 32.0 %. Size - 5L Storage Conditions - +15 °C TO +30 °C Catalog Number - 27570.02
Catalog No. 50-247-793