Learn More
Medchemexpress LLC Lauroylsarcosine sodium | 137-16-6 | 99.7% | 293.38 g/mol | C15H28NNaO3 | 10MM 1ML
SDP
Catalog No. 5000237407
Encompass_Preferred
Lauroylsarcosine sodium is the sodium salt of lauroylsarcosine, a surfactant reagent used to lower surface tension and aid solubilization in formulation and biochemical research. It is supplied as a ready-to-use 10 mM solution (1 mL) or as a bulk solid and is accompanied by analytical documentation to support research use.
  • Ready-to-use 10 mM solution, 1 mL
  • High purity (~99.7%)
  • Soluble in water and DMSO; ultrasonic or gentle warming may be required
  • Available in multiple pack sizes for small-scale testing or bulk use
  • Supplier-provided COA, SDS, and analytical data available
Catalog No. 50-002-37407 Supplier Medchemexpress LLC Supplier No. HY12592010MM 1ML
Lauroylsarcosine sodium is the sodium salt of lauroylsarcosine, a surfactant reagent used to lower surface tension and aid solubilization in formulation and biochemical research. It is supplied as a ready-to-use 10 mM solution (1 mL) or as a bulk solid and is accompanied by analytical documentation to support research use.
  • Ready-to-use 10 mM solution, 1 mL
  • High purity (~99.7%)
  • Soluble in water and DMSO; ultrasonic or gentle warming may be required
  • Available in multiple pack sizes for small-scale testing or bulk use
  • Supplier-provided COA, SDS, and analytical data available