Learn More
Medchemexpress LLC Lauroylsarcosine sodium | 137-16-6 | MFCD00042728 | 99.7% | 293.38 g/mol | C15H28NNaO3 | 100G
SDP
Supplier:  Medchemexpress LLC HY125920100G
Encompass_Preferred
Lauroylsarcosine sodium is an anionic surfactant (sodium lauroylsarcosinate, CAS 137-16-6) commonly used in personal care formulations, detergents, and biochemical assays to lower surface tension and aid solubilization. It is supplied as a high-purity solid or solution for research and formulation purposes and is intended for research use only.
  • High purity (~99.7%) suitable for analytical and formulation use.
  • Anionic surfactant that provides gentle cleansing and foaming.
  • Effective emulsifier and solubilizer in aqueous systems.
  • Available in solid and solution formats for flexible dosing.
  • Supplied with CAS identification for unambiguous chemical identity.
Catalog No. 50-002-37406