Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lauryl acrylate

Catalog No. 447315100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-731-5100ML 100 mL
44-731-5500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-731-5100ML Supplier Sigma Aldrich Supplier No. 447315100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lauryl acrylate monomers are extremely hydrophobic in nature.Lauryl acrylate can undergo atom transfer radical polymerization (ATRP) to form its polymer. Lauryl acrylate may undergo photoinitiated polymerization in the presence of N-acetyl-4-nitro-1-naphthylamine (ANNA) and N,N-dimethylaniline (DMA).

Specifications

CAS 2156-97-0
Density 0.884 g/mL (at 25°C)
Molecular Formula C15H28O2
Linear Formula CH2=CHCOOC12H25
Refractive Index n20/D 1.445 (literature)
MDL Number MFCD00042871
Quantity 100 mL
Synonym Dodecyl acrylate
Molecular Weight (g/mol) 240.38
Percent Purity 90%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.