Learn More
Medchemexpress LLC Lauryl benzalkonium chloride | 139-07-1 | ≥97.0% | 100 MG
SDP
Catalog No. 5000411452
Encompass_Preferred
Lauryl benzalkonium chloride, also known as dimethylbenzyllaurylammonium chloride, is a cationic surfactant primarily intended for research use. It is supplied with a high purity and specific storage conditions to maintain stability.
  • Molecular formula C21H38ClN with a molecular weight of 340.00.
  • Appears as a white to light brown solid below 42°C and a liquid above 42°C.
  • Readily soluble in DMSO at a concentration of 100 mg/mL.
  • Recommended for sealed storage at 4°C, away from moisture.
  • For solutions, store at -80°C for up to 6 months or -20°C for 1 month.
Catalog No. 50-004-11452 Supplier Medchemexpress LLC Supplier No. HYB1549100MG
Lauryl benzalkonium chloride, also known as dimethylbenzyllaurylammonium chloride, is a cationic surfactant primarily intended for research use. It is supplied with a high purity and specific storage conditions to maintain stability.
  • Molecular formula C21H38ClN with a molecular weight of 340.00.
  • Appears as a white to light brown solid below 42°C and a liquid above 42°C.
  • Readily soluble in DMSO at a concentration of 100 mg/mL.
  • Recommended for sealed storage at 4°C, away from moisture.
  • For solutions, store at -80°C for up to 6 months or -20°C for 1 month.