Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lauryl benzalkonium chloride | 139-07-1 | ≥97.0% | 500 MG
SDP

Catalog No. 5000411493
Encompass_Preferred

Lauryl benzalkonium chloride (dimethylbenzyllaurylammonium chloride) is a cationic surfactant primarily used for research. This chemical is shipped at room temperature and requires specific storage conditions to maintain stability.

  • Cationic surfactant
  • For research use only
  • Ships at room temperature
  • Store at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
  • Appearance: White to light brown solid below 42°C, liquid above 42°C
  • Soluble in DMSO

Catalog No. 50-004-11493 Supplier Medchemexpress LLC Supplier No. HYB1549500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lauryl benzalkonium chloride (dimethylbenzyllaurylammonium chloride) is a cationic surfactant primarily used for research. This chemical is shipped at room temperature and requires specific storage conditions to maintain stability.

  • Cationic surfactant
  • For research use only
  • Ships at room temperature
  • Store at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
  • Appearance: White to light brown solid below 42°C, liquid above 42°C
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.