Learn More
Medchemexpress LLC Lauryl glucoside | 110615-47-9 | MFCD19980983 | 50.0% | 348.47 g/mol | C18H36O6 | 10 MG
SDP
Catalog No. 5000285231
Encompass_Preferred
Lauryl glucoside (50% in water) is a non-ionic surfactant provided as a 50% w/w aqueous solution. It delivers high cleansing and foaming performance and is supplied as a viscous, colorless to light-yellow liquid for laboratory research applications. Refer to the safety data sheet for handling and storage details.
  • Non-ionic surfactant with high cleansing and foaming ability
  • Supplied as a 50% aqueous formulation
  • Viscous liquid, colorless to light yellow
  • Contains substance identified by CAS 110615-47-9
  • For research use only; follow SDS for handling and storage
Catalog No. 50-002-85231 Supplier Medchemexpress LLC Supplier No. HYW39217310ML
Lauryl glucoside (50% in water) is a non-ionic surfactant provided as a 50% w/w aqueous solution. It delivers high cleansing and foaming performance and is supplied as a viscous, colorless to light-yellow liquid for laboratory research applications. Refer to the safety data sheet for handling and storage details.
  • Non-ionic surfactant with high cleansing and foaming ability
  • Supplied as a 50% aqueous formulation
  • Viscous liquid, colorless to light yellow
  • Contains substance identified by CAS 110615-47-9
  • For research use only; follow SDS for handling and storage