Learn More
Medchemexpress LLC Lauryl glucoside | 110615-47-9 | MFCD19980983 | 50.0% | 348.48 g/mol | C18H36O6 | 100 MG
SDP
Catalog No. 5000285287
Encompass_Preferred
Lauryl glucoside is a mild, biodegradable non-ionic surfactant supplied as a 50% aqueous solution. It offers high cleansing and foaming performance with low irritation potential, making it suitable for rinse-off and gentle leave-on formulations in personal care and household products.
  • Mild cleansing and foaming action.
  • Low irritation potential for sensitive skin applications.
  • Biodegradable and environmentally friendly.
  • Suitable for shampoos, body washes, and cosmetic formulations.
  • Supplied as a 50% aqueous solution for easy handling and dosing.
  • Cost-effective surfactant for formulation use.
Catalog No. 50-002-85287 Supplier Medchemexpress LLC Supplier No. HYW392173100ML
Lauryl glucoside is a mild, biodegradable non-ionic surfactant supplied as a 50% aqueous solution. It offers high cleansing and foaming performance with low irritation potential, making it suitable for rinse-off and gentle leave-on formulations in personal care and household products.
  • Mild cleansing and foaming action.
  • Low irritation potential for sensitive skin applications.
  • Biodegradable and environmentally friendly.
  • Suitable for shampoos, body washes, and cosmetic formulations.
  • Supplied as a 50% aqueous solution for easy handling and dosing.
  • Cost-effective surfactant for formulation use.