Learn More
Medchemexpress LLC Lauryl glucoside | 110615-47-9 | MFCD19980983 | 50.0% | 348.5 g/mol | C18H36O6 | 50 MG
SDP
Catalog No. 5000285172
Encompass_Preferred
Lauryl glucoside (50% in water) is a non-ionic surfactant supplied as a 50% w/w aqueous solution. It provides mild, effective cleansing and good foaming performance for formulation and laboratory research. The material is a viscous, colorless to light yellow liquid and analytical documentation (COA) and safety data (SDS) are available from the manufacturer.
Catalog No. 50-002-85172 Supplier Medchemexpress LLC Supplier No. HYW39217350ML
Lauryl glucoside (50% in water) is a non-ionic surfactant supplied as a 50% w/w aqueous solution. It provides mild, effective cleansing and good foaming performance for formulation and laboratory research. The material is a viscous, colorless to light yellow liquid and analytical documentation (COA) and safety data (SDS) are available from the manufacturer.