Learn More
Medchemexpress LLC Lauryl glucoside | 110615-47-9 | MFCD19980983 | 50.0% | 348.5 g/mol | C18H36O6 | 500 MG
SDP
Catalog No. 5000285202
Encompass_Preferred
Lauryl glucoside (50% in water) is a non-ionic surfactant supplied as a 50% w/w aqueous solution. It delivers effective cleansing and high foaming performance while maintaining mild skin compatibility, making it suitable for research and formulation applications requiring a gentle, water-based surfactant.
  • Non-ionic surfactant with high cleansing ability
  • High foaming capacity for effective formulation performance
  • Mild and biodegradable, suitable for gentle applications
  • Supplied as a 50% aqueous solution for easier handling and dosing
  • Useful for cleansing, emulsifying, and foaming formulations
Catalog No. 50-002-85202 Supplier Medchemexpress LLC Supplier No. HYW392173500ML
Lauryl glucoside (50% in water) is a non-ionic surfactant supplied as a 50% w/w aqueous solution. It delivers effective cleansing and high foaming performance while maintaining mild skin compatibility, making it suitable for research and formulation applications requiring a gentle, water-based surfactant.
  • Non-ionic surfactant with high cleansing ability
  • High foaming capacity for effective formulation performance
  • Mild and biodegradable, suitable for gentle applications
  • Supplied as a 50% aqueous solution for easier handling and dosing
  • Useful for cleansing, emulsifying, and foaming formulations