Learn More
Medchemexpress LLC Lauryl laurate | 13945-76-1 | MFCD00056150 | >99.0% | 368.64 g/mol | C24H48O2 | 25 G
SDP
Catalog No. 5000284975
Encompass_Preferred
Lauryl laurate (dodecyl dodecanoate) is a long-chain fatty acid ester used as an emollient, lubricant, and biochemical reagent. It is oil-soluble and commonly used as an intermediate and formulation ingredient in cosmetics and laboratory applications.
  • Long-chain fatty ester with emollient properties.
  • Oil-soluble and compatible with anhydrous formulations.
  • Waxy solid or low-melting oil-like appearance at ambient temperatures.
  • Used as an intermediate and formulation ingredient in cosmetics and laboratory reagents.
  • Molecular weight 368.64 g/mol and formula C24H48O2.
Catalog No. 50-002-84975 Supplier Medchemexpress LLC Supplier No. HYW61444325G
Lauryl laurate (dodecyl dodecanoate) is a long-chain fatty acid ester used as an emollient, lubricant, and biochemical reagent. It is oil-soluble and commonly used as an intermediate and formulation ingredient in cosmetics and laboratory applications.
  • Long-chain fatty ester with emollient properties.
  • Oil-soluble and compatible with anhydrous formulations.
  • Waxy solid or low-melting oil-like appearance at ambient temperatures.
  • Used as an intermediate and formulation ingredient in cosmetics and laboratory reagents.
  • Molecular weight 368.64 g/mol and formula C24H48O2.