Learn More
Apexbio Technology LLC Lauryl maltose neopentyl glycol 1257852-96-2 500mg
SDP
Catalog No. 5000093976
Encompass_Preferred
Lauryl maltose neopentyl glycol (LMNG CAS 1257852-96-2) is a nonionic detergent widely utilized in membrane protein research Its chemical structure promotes efficient solubilization and stabilization of membrane proteins facilitating extraction from biological membranes while preserving native conformation and functional activity Compared to conventional detergents LMNG enables membrane protein extraction at lower concentrations and displays reduced destabilizing effects on target proteins Its favorable biocompatibility and low toxicity make it particularly suitable for structural and functional studies of membrane proteins including X-ray crystallography and cryo-EM applications
Catalog No. 50-000-93976 Supplier Apexbio Technology LLC Supplier No. A9951500
Lauryl maltose neopentyl glycol (LMNG CAS 1257852-96-2) is a nonionic detergent widely utilized in membrane protein research Its chemical structure promotes efficient solubilization and stabilization of membrane proteins facilitating extraction from biological membranes while preserving native conformation and functional activity Compared to conventional detergents LMNG enables membrane protein extraction at lower concentrations and displays reduced destabilizing effects on target proteins Its favorable biocompatibility and low toxicity make it particularly suitable for structural and functional studies of membrane proteins including X-ray crystallography and cryo-EM applications