Learn More
Bioworld Lauryl Sulfate Broth, 500 g
Catalog No. 50254310
Encompass_Preferred
Lauryl Sulfate BrothFor the detection of coliforms in water, wastewater and foodspH: 6.8Working Concentration: 35.6 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: tan to yellow brothSoluble in waterComposition:Pancreatic Digest of Casein Lactose Sodium Chloride Monopotassium Phosphate Dipotassium Phosphate Sodium Lauryl Sulfate
Catalog No. 50-254-310 Supplier Bioworld Supplier No. 306200341
Lauryl Sulfate BrothFor the detection of coliforms in water, wastewater and foodspH: 6.8Working Concentration: 35.6 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: tan to yellow brothSoluble in waterComposition:Pancreatic Digest of Casein Lactose Sodium Chloride Monopotassium Phosphate Dipotassium Phosphate Sodium Lauryl Sulfate