Learn More
NEOGEN Lauryl Sulfate Broth with MUG, 10 kg
SDP
Catalog No. 502015362
Encompass_Preferred
Lauryl Sulfate Broth with MUG is used for the detection of coliforms and the fluorogenic detection of Escherichia coli in a laboratory setting. Lauryl Sulfate Broth with MUG is not intended for use in the diagnosis of disease or other conditions in humans.The coliform group of bacteria includes aerobic and facultative anaerobic, Gram-negative, non-sporeforming bacilli that ferment lactose and form acid and gas at 35°C within 48 hours. Members of the Enterobacteriacae comprise the majority of this group, but organisms such as Aeromonas spp. may also be included. Procedures to detect and confirm coliforms are used in testing water, foods, dairy products, and other materials.Lauryl Sulfate Broth, also referred to as Lauryl Tryptose Broth, is prepared according to the formula of Mallmann and Darby.For more information visit https://www.neogen.com/solutions/Microbiology/lauryl-sulfate-broth-with-mug
Catalog No. 50-201-5362 Supplier NEOGEN Supplier No. 700003171
Add to Cart
Add to Cart
Request a Quote
Lauryl Sulfate Broth with MUG is used for the detection of coliforms and the fluorogenic detection of Escherichia coli in a laboratory setting. Lauryl Sulfate Broth with MUG is not intended for use in the diagnosis of disease or other conditions in humans.The coliform group of bacteria includes aerobic and facultative anaerobic, Gram-negative, non-sporeforming bacilli that ferment lactose and form acid and gas at 35°C within 48 hours. Members of the Enterobacteriacae comprise the majority of this group, but organisms such as Aeromonas spp. may also be included. Procedures to detect and confirm coliforms are used in testing water, foods, dairy products, and other materials.Lauryl Sulfate Broth, also referred to as Lauryl Tryptose Broth, is prepared according to the formula of Mallmann and Darby.For more information visit https://www.neogen.com/solutions/Microbiology/lauryl-sulfate-broth-with-mug