Learn More
Alkali Scientific Lauryl Sulfate Tryptose Broth 10mL, 10/Pk
SDP
Catalog No. 5000663147
Encompass_Preferred
Lauryl Sulfate Tryptose (LST) Broth, 10mL Tubes, 10/Pk is a selective enrichment medium designed for the detection of coliform bacteria in water, food, and environmental samples. The formulation includes sodium lauryl sulfate, which inhibits the growth of non coliform organisms, while tryptose provides a nutrient rich base that supports the growth of Escherichia coli and related coliform species. Supplied in sterile 10mL tubes, the medium is suitable for Most Probable Number (MPN) testing and microbial limit analysis in quality control laboratories. The ready to use format simplifies preparation and maintains consistent testing conditions. LST broth is widely used in environmental monitoring, food safety testing, and microbiology laboratories for reliable detection and enumeration of coliform organisms.
Catalog No. 50-006-63147 Supplier Alkali Scientific Supplier No. LSTT00
Lauryl Sulfate Tryptose (LST) Broth, 10mL Tubes, 10/Pk is a selective enrichment medium designed for the detection of coliform bacteria in water, food, and environmental samples. The formulation includes sodium lauryl sulfate, which inhibits the growth of non coliform organisms, while tryptose provides a nutrient rich base that supports the growth of Escherichia coli and related coliform species. Supplied in sterile 10mL tubes, the medium is suitable for Most Probable Number (MPN) testing and microbial limit analysis in quality control laboratories. The ready to use format simplifies preparation and maintains consistent testing conditions. LST broth is widely used in environmental monitoring, food safety testing, and microbiology laboratories for reliable detection and enumeration of coliform organisms.