Learn More
HIMEDIA Lauryl Sulphate Broth (Lauryl Tryptose Broth), M080, Microbiology Culture Media, Dehydrated Culture Media, Broth, Coliform Detection, ISO 11133, Water and Food, Powder, 500 g
Catalog No. NC4013663
Encompass_Preferred
Lauryl Sulphate Broth, also known as Lauryl Tryptose Broth, detects and enumerates coliform bacteria in water, wastewater, dairy and other foods, with composition and performance meeting ISO 11133 (2014 and Amd. 2018). Tryptose and lactose nourish and reveal coliforms while a phosphate buffer stabilizes pH and sodium lauryl sulphate suppresses non-coliforms. Gas trapped in an inverted Durham tube signals a positive presumptive result. A standard presumptive coliform medium. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013663 Supplier HIMEDIA Supplier No. M080500G
Lauryl Sulphate Broth, also known as Lauryl Tryptose Broth, detects and enumerates coliform bacteria in water, wastewater, dairy and other foods, with composition and performance meeting ISO 11133 (2014 and Amd. 2018). Tryptose and lactose nourish and reveal coliforms while a phosphate buffer stabilizes pH and sodium lauryl sulphate suppresses non-coliforms. Gas trapped in an inverted Durham tube signals a positive presumptive result. A standard presumptive coliform medium. Dehydrated powder, store in a cool dry place. Packaged as 500 g.