Learn More
NEOGEN Lauryl Tryptose (LST) Broth, 10 kg
SDP
Catalog No. 502015248
Encompass_Preferred
Lauryl Tryptose (LST) Broth is used for the detection of coliform bacteria in water and wastewater. Lauryl Tryptose Broth is not intended for use in the diagnosis of disease or other conditions in humans.Lauryl Tryptose Broth is a selective medium for the detection of coliforms in water, dairy products and other foods. The American Public Health Authority (APHA) recommend Lauryl Tryptose Broth for the Most Probable Number Presumptive Test of coliforms in waters, effluent or sewage and as a confirmation test of lactose fermentation with gas production from milk samples and for the detection of coliforms in foods.Lauryl Tryptose Broth is prepared according to the formulation of Mallmann and Darby. Mallmann and Darby showed that tryptose at a concentration of 2% increased the early logarithmic growth phase when compared to meat peptone.For more information visit https://www.neogen.com/solutions/Microbiology/lauryl-tryptose-broth
Catalog No. 50-201-5248 Supplier NEOGEN Supplier No. 700003059
Lauryl Tryptose (LST) Broth is used for the detection of coliform bacteria in water and wastewater. Lauryl Tryptose Broth is not intended for use in the diagnosis of disease or other conditions in humans.Lauryl Tryptose Broth is a selective medium for the detection of coliforms in water, dairy products and other foods. The American Public Health Authority (APHA) recommend Lauryl Tryptose Broth for the Most Probable Number Presumptive Test of coliforms in waters, effluent or sewage and as a confirmation test of lactose fermentation with gas production from milk samples and for the detection of coliforms in foods.Lauryl Tryptose Broth is prepared according to the formulation of Mallmann and Darby. Mallmann and Darby showed that tryptose at a concentration of 2% increased the early logarithmic growth phase when compared to meat peptone.For more information visit https://www.neogen.com/solutions/Microbiology/lauryl-tryptose-broth