Learn More
Alkali Scientific Lauryl Tryptose Broth, 2 Kilograms
SDP
Catalog No. 5000659745
Encompass_Preferred
Lauryl Tryptose Broth, 2 Kilograms, is a selective broth medium used for the growth and differentiation of coliform bacteria in microbiological testing. Ideal for laboratory applications, this medium supports the growth of a broad range of microorganisms, including fecal coliforms, making it highly valuable for water and food quality testing. This quality-controlled product ensures accurate, reliable results in all testing scenarios.
Catalog No. 50-006-59745 Supplier Alkali Scientific Supplier No. LAU003CP
Lauryl Tryptose Broth, 2 Kilograms, is a selective broth medium used for the growth and differentiation of coliform bacteria in microbiological testing. Ideal for laboratory applications, this medium supports the growth of a broad range of microorganisms, including fecal coliforms, making it highly valuable for water and food quality testing. This quality-controlled product ensures accurate, reliable results in all testing scenarios.