Learn More
Hach Company Lauryl Tryptose Broth, Dehydrated 500 g
Supplier:  Hach Company 19734
Encompass
Lauryl tryptose broth is used to detect coliform bacteria in water and wastewater using the Most Probable Number (MPN) method.
Catalog No. 50-199-4199