Learn More
Hach Company Lauryl Tryptose Broth MPN Tubes, Concentrated, pk/15
Supplier:  Hach Company 2101415
Encompass
Lauryl Tryptose Broth MPN Tubes, concentrated, pk/15. Selectivity: Total Coliform Bacteria (presumptive).
Catalog No. 50-199-5386