Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lavandulyl acetate | 25905-14-0 | MFCD16652611 | 98.0% | 196.29 g/mol | C12H20O2 | 10 MG
SDP

Catalog No. 5000228646
Encompass_Preferred

(±)-Lavandulyl acetate is the racemic form of lavandulyl acetate, a naturally occurring monoterpenoid ester isolated from Lavandula angustifolia (lavender). It is supplied as a high-purity, colorless to light yellow liquid for research use, suitable for analytical reference standards, olfactory studies, and natural product research. For research use only; not for human consumption.

  • High purity (98.0%).
  • Molecular weight 196.29 g/mol.
  • CAS number 25905-14-0.
  • Liquid appearance, colorless to light yellow.
  • Soluble in DMSO.
  • Available in small quantities suitable for analytical work.

Catalog No. 50-002-28646 Supplier Medchemexpress LLC Supplier No. HY117419A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-Lavandulyl acetate is the racemic form of lavandulyl acetate, a naturally occurring monoterpenoid ester isolated from Lavandula angustifolia (lavender). It is supplied as a high-purity, colorless to light yellow liquid for research use, suitable for analytical reference standards, olfactory studies, and natural product research. For research use only; not for human consumption.

  • High purity (98.0%).
  • Molecular weight 196.29 g/mol.
  • CAS number 25905-14-0.
  • Liquid appearance, colorless to light yellow.
  • Soluble in DMSO.
  • Available in small quantities suitable for analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.