Learn More
Medchemexpress LLC Lavendustin A | 125697-92-9 | 99.0% | 381.38 | 1 MG
SDP
Catalog No. 5000385110
Encompass_Preferred
Lavendustin A (RG-14355) is a potent, selective, and ATP-competitive inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase, with an IC50 of 11 nM. It does not inhibit protein kinase A or C. Lavendustin A can suppress VEGF-induced angiogenesis.
  • Potent, selective, and ATP-competitive inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase.
  • Does not inhibit protein kinase A or C.
  • Can suppress VEGF-induced angiogenesis.
Catalog No. 50-003-85110 Supplier Medchemexpress LLC Supplier No. HY189631MG
Lavendustin A (RG-14355) is a potent, selective, and ATP-competitive inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase, with an IC50 of 11 nM. It does not inhibit protein kinase A or C. Lavendustin A can suppress VEGF-induced angiogenesis.
  • Potent, selective, and ATP-competitive inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase.
  • Does not inhibit protein kinase A or C.
  • Can suppress VEGF-induced angiogenesis.