Learn More
Cayman Chemical Lavendustin A Inhibitors
SDP
Catalog No. 502890098
Encompass_Preferred
A selective inhibitor of EGF receptor-associated tyrosine kinase (IC50 = 11 nM); used to differentiate rat mesenchymal stem cells, to inhibit NMDA-stimulated cGMP production, and to inhibit VEGF-induced angiogenesis
Catalog No. 50-289-0098 Supplier Cayman Chemical Supplier No. 100102685 MG
A selective inhibitor of EGF receptor-associated tyrosine kinase (IC50 = 11 nM); used to differentiate rat mesenchymal stem cells, to inhibit NMDA-stimulated cGMP production, and to inhibit VEGF-induced angiogenesis