Learn More
Cayman Chemical LavendustIn B 10mg
SDP
Catalog No. 502939862
Encompass_Preferred
An inhibitor of Glut1 (Ki = 15 µM); an inhibitor of the interaction between HIV-1 integrase and LEDGF/p75 (IC50 = 94.07 µM); a weak inhibitor of tyrosine kinases (IC50 = 0.49 UG/ml); and has been used as a negative control for lavendustin A
Catalog No. 50-293-9862 Supplier Cayman Chemical Supplier No. 2731110 MG
An inhibitor of Glut1 (Ki = 15 µM); an inhibitor of the interaction between HIV-1 integrase and LEDGF/p75 (IC50 = 94.07 µM); a weak inhibitor of tyrosine kinases (IC50 = 0.49 UG/ml); and has been used as a negative control for lavendustin A